TriLink® Pseudouridine-5'-Triphosphate
Leverandør: TriLink BioTechnologies
N-1019-10 N-1019-1 N-1019-5
SOLUN-1019-10EA 6075 NOK
SOLUN-1019-10 SOLUN-1019-1 SOLUN-1019-5
TriLink® Pseudouridine-5'-Triphosphate
Nukleinsyre reagenser Nukleotider
This nucleotide consists of a modified base, a ribose, and a 5′ triphosphate group.
  • Can be incorporated in in vitro transcription for modification of synthesised RNA for desired characteristics
  • ≥97% purity by AX-HPLC analysis, provided as 100 mM in H₂O
  • Grade: RUO
Pseudouridine has been found to enhance base stacking and translation. Pseudouridine-5'-triphosphate (Pseudo-UTP) is used to impart desirable mRNA characteristics such as increased nuclease stability.
NB: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=92efad7a00d4ca61a780f899f28d0a583a797b53&vid=92efad7a00d4ca61a780f899f28d0a583a797b53&prid=web&clid=SAPDAM
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Applikasjon