TriLink® 2'-Deoxyguanosine-5'-Triphosphate
Leverandør: TriLink BioTechnologies
N-2503-100 N-2503-25
SOLUN-2503-100EA 2675 NOK
SOLUN-2503-100 SOLUN-2503-25
TriLink® 2'-Deoxyguanosine-5'-Triphosphate
Nukleinsyre reagenser Nukleotider
This nucleotide consists of an unmodified base, a deoxyribose, and a 5′ triphosphate group for synthesis of DNA molecules.
  • Unmodified dexoyribonucleotide for DNA synthesis
  • ≥97% purity by AX-HPLC analysis
  • Provided as 100 mM in H₂O, pH 7,5
  • Grade: RUO
Deoxyadenosine-5′-triphosphate (dATP), deoxycytidine-5′-triphosphate (dCTP), deoxyguanosine-5′-triphosphate (dGTP), and deoxythymidine-5′-triphosphate (dTTP) are unmodified deoxyribonucleoside triphosphates, serving as essential building blocks of DNA molecules. Each of them consists of an unmodified base, a dexoyribose, and a 5′ triphosphate group.
NB: Item will be delivered at or below −20 °C and should be stored at or below −20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cadaed7ca0f3b918abf818cdafbc2b4a23897725&vid=cadaed7ca0f3b918abf818cdafbc2b4a23897725&prid=web&clid=SAPDAM
New Product
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Applikasjon