Eyel model
Leverandør: SOMMER MARCUS
CS10
SOMMCS10EA 0 NOK
SOMMCS10
Eyel model
Modeller Human biologi
Resting in the bone of the base of the orbit, median section. In the left half the lens and vitreous humour are fixed. The right half shows the sclerotic membrane partially opened from the outside in order to expose the retina. The interior side shows the ciliary body and the background of the eye. A section of the retina shows the structural scheme of the choroid membrane with retina:
  • Enlarged approximately five times
  • On a green base with explanation
  • Separates altogether in three parts