Worktop trolley, ESD
Leverandør: TRESTON
C70135001
SOVEC70135001EA 0 NOK
SOVEC70135001
Worktop trolley, ESD
Traller
Worktop trolley for workspace with ESD-protection.
  • Height adjustable from 655 to 1055 mm
  • Four swivel wheels (Ø 100), two with brakes
Worktop and bottom shelf laminated ESD surface with beech edges.
Bestillingsinformasjon: Also available as non-ESD.