Pectin (from apple), powder

Leverandør: Spectrum Chemical

A1201-2.5KG
SPCMA1201-2.5KGEA 5350 NOK
SPCMA1201-2.5KG
Pectin (from apple), powder
Pectin
MDL nummer: MFCD00081838
CAS-nummer: 9000-69-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR