Diammoniumhydrogenortofosfat, granular NF

Leverandør: Spectrum Chemical

A1244-12KG A1244-2.5KG A1244-45KGBL
SPCMA1244-12KGEA 21375 NOK
SPCMA1244-12KG SPCMA1244-2.5KG SPCMA1244-45KGBL
Diammoniumhydrogenortofosfat, granular NF
Diammoniumhydrogenortofosfat

Melamine statement (available upon request)
BSE/TSE statement (available upon request)

Formel: (NH₄)₂HPO₄
Smeltepunkt: 185 °C
Tetthet: 1,619 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010891
CAS-nummer: 7783-28-0
EINECS: 231-987-8
REACH: 01-2119490974-22
Merck Indeks: 13,00546

Bestill nå

Spesifikasjon Testresultater

Assay 96% Min.

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR