Adenin (synthetic)

Leverandør: Spectrum Chemical
Danger

AD105-1KGBL
SPCMAD105-1KGBLEA 59475 NOK
SPCMAD105-1KGBL
Adenin (synthetic)
Adenin
Formel: C₅H₅N₅
Tetthet: 0,99741 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00041790
CAS-nummer: 73-24-5
EINECS: 200-796-1
FN: 2811
ADR: 6.1,III
Merck Indeks: 13,00152

Bestill nå

Spesifikasjon Testresultater

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR