L(+)-Askorbinsyre ≥99.0% USP

Leverandør: Spectrum Chemical

AS102-25GM
SPCMAS102-25GMEA 1400 NOK
SPCMAS102-25GM
L(+)-Askorbinsyre ≥99.0% USP
L(+)-Askorbinsyre

Spectrum-labeled kosher products with the Kosher Supervision of America symbol are a firm guarantee to all Spectrum customers that the products bearing the symbol are in full compliance with the most demanding of Kosher and Pareve standards.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: C₆H₈O₆
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064328
CAS-nummer: 50-81-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c03499908e269560d680b0d7c4651f97a367aa0&vid=73a720d6900e713a98164022273a8afa6114ad7b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay 99.0 - 100.5 %
Specific Rotation [A]D +20.5 to +21.5
Residue On Ignition max. 0.1 %
Heavy Metals (As Pb) max. 20 ppm
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR