Betaine, anhydrous

Leverandør: Spectrum Chemical

Synonymer: TMG

B1572-25KGBL
SPCMB1572-25KGBLEA 0 NOK
SPCMB1572-25KGBL
Betaine, anhydrous
Betaine
Formel: C₅H₁₁NO₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012123
CAS-nummer: 107-43-7
EINECS: 203-490-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR