Brij® O2

Leverandør: Spectrum Chemical

B1676-200LTBL
SPCMB1676-200LTBLEA 285850 NOK
SPCMB1676-200LTBL
Brij® O2
Brij® O2
MDL nummer: MFCD00074369
CAS-nummer: 9004-98-2
EINECS: 500-016-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR