2-Butyne-1,4-diol ≥99%

Leverandør: Spectrum Chemical

B2232-50KGBL
SPCMB2232-50KGBLEA 131925 NOK
SPCMB2232-50KGBL
2-Butyne-1,4-diol ≥99%
2-Butyne-1,4-diol

2-Butyne-1,4-diol is a colorless, hygroscopic solid that is soluble in water and polar organic solvents. It is primarily used as a synthetic organic intermediate used in the manufacture of certain herbicides, textile additives, corrosion inhibitors, plasticizers, synthetic resins, and polyurethanes. It is the major raw material of vitamin B6 and is also used for brightening, preserving, and inhibiting nickel plating.


GHS hazard : Skull and crossbones;Exclamation mark;Health hazard
UN2716

Formel: C₄H₆O₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002915
CAS-nummer: 110-65-6
FN: 2716
ADR: 6.1,III

Bestill nå

Spesifikasjon Testresultater

Assay 99% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR