Brij® L23

Leverandør: Spectrum Chemical
Warning

BR105-12KG
SPCMBR105-12KGEA 34625 NOK
SPCMBR105-12KG
Brij® L23
Brij® L23

Brij(R) L23 is a non-ionic detergent for protein extraction and permeabilization of cells. It may be used in the preparation of yeast spheroplasts. It appears as a white, waxy solid.

Formel: (C₂₀H₄₂O₅)n
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00043063
CAS-nummer: 9002-92-0
EINECS: 500-002-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR