Carboxymethyl cellulose sodium salt FCC, low viscosity

Leverandør: Spectrum Chemical

C1103-2.5KG
SPCMC1103-2.5KGEA 10775 NOK
SPCMC1103-2.5KG
Carboxymethyl cellulose sodium salt FCC, low viscosity
Carboxymethyl cellulose sodium salt
Formel: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
Smeltepunkt: 300 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00081472
CAS-nummer: 9004-32-4
Merck Indeks: 12,01877

Bestill nå

Spesifikasjon Testresultater

Assay 99.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR