Carbomer 941 resin NF

Leverandør: Spectrum Chemical

C1478-2.5KG C1478-45KGBL C1478-100GM
SPCMC1478-2.5KGEA 13400 NOK
SPCMC1478-2.5KG SPCMC1478-45KGBL SPCMC1478-100GM
Carbomer 941 resin NF
Carbomer 941

Carbomer 941, NF is an expanded molecule that will increase continually when more alkyne bonds are presented.


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).

Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00084394
CAS-nummer: 9062-04-8

Bestill nå

Spesifikasjon Testresultater

Carboxylic Acid Groups (Dried Basis) 56.0 - 68.0%
Viscosity of a 0.5% Solution 4,000-11,000 mPa·s
Benzene max. 0.5%
Loss On Drying max. 2.0%
Heavy Metals max. 20 µg/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR