Cesium nitrate, Ultrapure

Leverandør: Spectrum Chemical

C1489-10KGBL C1489-1KG
SPCMC1489-10KGBLEA 147500 NOK
SPCMC1489-10KGBL SPCMC1489-1KG
Cesium nitrate, Ultrapure
Cesiumnitrat

Cesium Nitrate, Ultrapure is used as a colorant, as an oxidizer, and in pyrotechnic compositions.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: CsNO₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00010963
CAS-nummer: 7789-18-6
FN: 1451
ADR: 5.1,III

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR