Carmine

Leverandør: Spectrum Chemical

Synonymer: CI 75470

CA195-1KGBL CA195-5KGBL
SPCMCA195-1KGBLEA 70925 NOK
SPCMCA195-1KGBL SPCMCA195-5KGBL
Carmine
Carmine

Carmine, also known as cochineal or crimson lake, is a red colored pigment obtained from the aluminum salt of carminic acid. It is used in artificial flowers, paints and coloring. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₂₂H₂₀O₁₃
MW: 492.389 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00167028
CAS-nummer: 1390-65-4

Bestill nå

Spesifikasjon Testresultater

Absorption Max 531 (563) NM


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR