Dekstran, M.W. ~ 150,000, pulver

Leverandør: Spectrum Chemical

D1013-1KG
SPCMD1013-1KGEA 25500 NOK
SPCMD1013-1KG
Dekstran, M.W. ~ 150,000, pulver
Dekstran

Form: Powder

Formel: (C₆H₁₀O₅)n
MDL nummer: MFCD00130935
CAS-nummer: 9004-54-0
EINECS: 232-677-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR