Dimethyl glutarate ≥98%

Leverandør: Spectrum Chemical

D2948-200LTBL
SPCMD2948-200LTBLEA 68475 NOK
SPCMD2948-200LTBL
Dimethyl glutarate ≥98%
Dimethyl glutarate

Dimethyl Glutarate is used in de-icing and anti-icing applications, construction and road applications, and release and binders agents.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₇H₁₂O₄
Kokepunkt: 211 °C (1013 hPa)
Smeltepunkt: –37 °C
Tetthet: 1,087
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008468
CAS-nummer: 1119-40-0
EINECS: 214-277-2
Merck Indeks: 13,04486

Bestill nå

Spesifikasjon Testresultater

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR