Acid red 33

Leverandør: Spectrum Chemical

Synonymer: D&C Red No. 33, Disodium (3Z)-5-amino-4-oxo-3-(phenylhydrazinylidene)naphthalene-2,7-disulphonate, Azo Fuchsine

DC145-100GM DC145-25GM
SPCMDC145-100GMEA 9775 NOK
SPCMDC145-100GM SPCMDC145-25GM
Acid red 33
Acid red 33

D and C Red No. 33 | Non-graded D and C Red No. 33 supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₁₆H₁₁N₃Na₂O₇S₂
MW: 467.378 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00071877
CAS-nummer: 3567-66-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR