Quinoline yellow sulphonic acid sodium salt (Acid yellow 3)

Leverandør: Spectrum Chemical

DC200-25GM
SPCMDC200-25GMEA 2125 NOK
SPCMDC200-25GM
Quinoline yellow sulphonic acid sodium salt (Acid yellow 3)
Quinoline yellow sulphonic acid sodium salt (Acid yellow 3)

D and C Yellow No. 10 | Non-graded D and C Yellow No. 10 supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₁₈H₁₁NO₂.xNaO₃S
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00080727
CAS-nummer: 8004-92-0

Bestill nå

Spesifikasjon Testresultater

Arsenic (As) To Pass Test
Lead (Pb) To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR