Dextran, M.W. ~ 40,000, powder

Leverandør: Spectrum Chemical

DE124-1KG
SPCMDE124-1KGEA 17375 NOK
SPCMDE124-1KG
Dextran, M.W. ~ 40,000, powder
Dekstran

Dextran, 40,000, Powder is a complex branched glucan that is synthesized by certain lactic acid bacteria from sucrose. Commonly used in microsurgery to decrease the risk of vascular thrombosis, these agents are also used as effective osmotic agents and can treat hypovolemia. In the laboratory, Dextran is used to apply osmotic pressure to biological molecules and can, when applied with a fluorescent molecule, be used to create concentration gradients of diffusible molecules.

Formel: (C₆H₁₀O₅)n
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00130935
CAS-nummer: 9004-54-0
EINECS: 232-677-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR