Iron(II) sulphate hydrate, dried powder FCC

Leverandør: Spectrum Chemical

F1063-500GM
SPCMF1063-500GMEA 3250 NOK
SPCMF1063-500GM
Iron(II) sulphate hydrate, dried powder FCC
Iron(II) sulphate hydrate

Ferrous Sulfate, Dried Powder, FCC is used as an iron fortification food supplement additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formel: FeSO₄·H₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00149722
CAS-nummer: 13463-43-9

Bestill nå

Spesifikasjon Testresultater

Assay (FeSO₄) 86.0 - 89.0%
Insoluble Substances max. 0.05 %
Lead (Pb) max. 2 mg/kg
Mercury max. 1 mg/kg
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR