Indigo carmine

Leverandør: Spectrum Chemical
Warning

FD115-100GM FD115-25GM FD115-500GM
SPCMFD115-100GMEA 2375 NOK
SPCMFD115-100GM SPCMFD115-25GM SPCMFD115-500GM
Indigo carmine
Indigo carmine

FD and C Blue No. 2 - In the United States, the Food and Drug Administration (FDA) controls the specifications for Food, Drug and Cosmetic (FD&C) colours.


Spectrum certifies all batches of FD&C colours meet the FDA's quality standards.


NB: Storage temperature - LIGHT SENSITIVE: Keep tightly closed in light-resistant containers.

Formel: C₁₆H₈N₂Na₂O₈S₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005723
CAS-nummer: 860-22-0

Bestill nå

Spesifikasjon Testresultater

Identification To Pass Test

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR