Folic acid ≥97.0%, powder USP

Leverandør: Spectrum Chemical

Synonymer: Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9

FO105-1KG FO105-25KGBL FO105-5KGBL FO105-100GM
SPCMFO105-1KGEA 56650 NOK
SPCMFO105-1KG SPCMFO105-25KGBL SPCMFO105-5KGBL SPCMFO105-100GM
Folic acid ≥97.0%, powder USP
Folic acid

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: C₁₉H₁₉N₇O₆
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00079305
CAS-nummer: 59-30-3

Bestill nå

Spesifikasjon Testresultater

Assay (Anhydrous Basis) 97.0 - 102.0%
Water Determination max.8.5 %
Residue On Ignition max. 0.3 %
Related Compounds max. 2.0 %
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR