Magnesium hydroxide ≥95%, pulver USP

Leverandør: Spectrum Chemical

MA115-2.5KG
SPCMMA115-2.5KGEA 4600 NOK
SPCMMA115-2.5KG
Magnesium hydroxide ≥95%, pulver USP
Magnesiumhydroksid

Magnesium Hydroxide, Powder, USP is used to treat symptoms caused by too much stomach acid such as heartburn, upset stomach, or indigestion. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: Mg(OH)₂
Tetthet: 2,3261…2,3631 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011104
CAS-nummer: 1309-42-8
EINECS: 215-170-3
Merck Indeks: 13,05693

Bestill nå

Spesifikasjon Testresultater

Assay 95% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR