Potassium sorbate ≥98.0%, pulver FCC

Leverandør: Spectrum Chemical

Synonymer: Potassium 2,4-Hexadienoate, Sorbic acid potassium salt, Potassium (2E,4E)-hexa-2,4-dienoate

P1408-125GM
SPCMP1408-125GMEA 1725 NOK
SPCMP1408-125GM
Potassium sorbate ≥98.0%, pulver FCC
Kaliumsorbat

Potassium Sorbate, Powder, FCC is used as an antimicrobial preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplement and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formel: CH₃CH=CHCH=CHCOOK
MW: 150,22 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00016546
CAS-nummer: 24634-61-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=500421971592bd8f37c1a6b88f97c1de033cbf87&vid=e8c4a1d0da78946f174af6a94e5c78fc9c16de05&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (Dried Basis) 98.0 - 101.0%
Acidity To Pass Test
Alkalinity To Pass Test
Lead (Pb) max. 2 mg/kg
Loss On Drying max. 1.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR