Propansyre 99.5-100.5% NF

Leverandør: Spectrum Chemical
Danger

P1464-200LTBL P1464-20LTBL P1464-4LTGL
SPCMP1464-200LTBLEA 560500 NOK
SPCMP1464-200LTBL SPCMP1464-20LTBL SPCMP1464-4LTGL
Propansyre 99.5-100.5% NF
Propansyre

Propionic acid is used as a preservative in pharmaceutical formulations.


Sertifiseringer: Manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: C₃H₆O₂
Kokepunkt: 141 °C (1013 hPa)
Smeltepunkt: –21 °C
Tetthet: 0,99 g/cm³ (20 °C)
Flash punkt: 49 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002756
CAS-nummer: 79-09-4
EINECS: 201-176-3
FN: 3463
ADR: 8,II
REACH: 01-2119486971-24
Merck Indeks: 13,07917

Bestill nå

Spesifikasjon Testresultater

Assay Min. 99.5 - 100.5%
Specific gravity at 25 °C 0.988 - 0.993
Distilling Range 138.5 - 142.5 °C
Nonvolatile Residue 0.0001
Elemental Impurities As Reported
Readily Oxidizable Substances To Pass Test
Aldehyde To Pass Test
Identification To Pass Test
Residual Solvents To Pass Test

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR