Potassium iodide ≥99.0% USP

Leverandør: Spectrum Chemical

PO185-125GM
SPCMPO185-125GMEA 2825 NOK
SPCMPO185-125GM
Potassium iodide ≥99.0% USP
Kaliumjodid

Spectrum-labeled kosher products with the Kosher Supervision of America symbol are a firm guarantee to all Spectrum customers that the products bearing the symbol are in full compliance with the most demanding of Kosher and Pareve standards.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: Kl
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011405
CAS-nummer: 7681-11-0

Bestill nå

Spesifikasjon Testresultater

Assay 99.0 - 101.5%
Alkalinity To Pass Test
Loss On Drying max. 1.0 %
Iodate max. 4 Ug/G
Nitrate, Nitrite, and Ammonia To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR