Sølvnitrat ≥99.8%, krystaller USP

Leverandør: Spectrum Chemical
Danger

SI115-100GM SI115-500GM
SPCMSI115-100GMEA 31425 NOK
SPCMSI115-100GM SPCMSI115-500GM
Sølvnitrat ≥99.8%, krystaller USP
Sølvnitrat

Silver Nitrate, Crystal, USP is used as an antibiotic for indolent wounds, corn and callus removal and as a Gonorrheal ophthalmia neonatorum prophylaxis. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: AgNO₃
Kokepunkt: 444 °C (1013 hPa)
Smeltepunkt: 210 °C
Tetthet: 4,35 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003414
CAS-nummer: 7761-88-8
EINECS: 231-853-9
FN: 1493
ADR: 5.1,II
REACH: 01-2119513705-43
Merck Indeks: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay 99.8% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR