tri-Sodium citrate dihydrate ≥99.0%, kornet USP

Leverandør: Spectrum Chemical

SO165-500GM
SPCMSO165-500GMEA 2075 NOK
SPCMSO165-500GM
tri-Sodium citrate dihydrate ≥99.0%, kornet USP
tri-Natriumcitrat dihydrat

Sodium Citrate, Dihydrate, Granular, USP can be used as an alkalising agent, buffering agent, emulsifier, or sequestering agent.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: Na₃C₆H₅O₇·2H₂O
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00150031
CAS-nummer: 6132-04-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Assay (C₆H₅Na₃O₇) 99.0 - 100.5%
Water 10.0 - 13.0 %
Alkalinity To Pass Test
Tartrate To Pass Test
Heavy Metals max. 0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR