L-Threonine ≥98.5%, krystaller USP

Leverandør: Spectrum Chemical

T1056-25GM
SPCMT1056-25GMEA 1725 NOK
SPCMT1056-25GM
L-Threonine ≥98.5%, krystaller USP
L-Threonine

Threonine, Crystal, USP is an amino acid that is used in the biosynthesis of proteins. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: C₄H₉NO₃
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064270
CAS-nummer: 72-19-5
EINECS: 200-774-1

Bestill nå

Spesifikasjon Testresultater

Assay 98.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR