Zinc picolinate, pulver 200-400 Mesh

Leverandør: Spectrum Chemical

Z1088-25KGBL
SPCMZ1088-25KGBLEA 132175 NOK
SPCMZ1088-25KGBL
Zinc picolinate, pulver 200-400 Mesh
Zinc picolinate

Zinc Picolinate, 200 to 400 Mesh, Powder is most commonly used as a nutritional food supplement as a source for zinc and aspartic acid.

Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00145544
CAS-nummer: 17949-65-4

Bestill nå

Spesifikasjon Testresultater

Assay Aprx.200 MG Zn/G
Lead (Pb) max. 10 ppm
Arsenic (As) max. 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR