KR2i Starter Fitting Kit
Leverandør: Spectrum Laboratories
ACR2-SKT
SPECACR2-SKTEA 39075 NOK
SPECACR2-SKT
KR2i Starter Fitting Kit
Fittings og kontakter
Kit with Luer, mini TC accessories and tools.
Kit consists of fittings: 3 Luer pressure Sensor, non-irradiated (PS), 5 flow control valve (PS), 20 orange cable tie 152,4L×9,52 W mm one way stopcock (nylon), 4 sanitary clamp 12,7×19,05 mm(nylon), 10 sanitary gakset 19,05 mm (silicone), 4 ProConnex®, 19,05 mm TC×6,35 mm HB with FLL (PS), 5 stopcock (PS), 6 ProConnex®, 12,7 mm TC to FLL (PS), 6 Sanitary gasket 12,7 mm (silicone).
Consists of tools: 1 cable tie gun, 1 scissors, 2 cable box and 1 screwdriver (Philips).