Hollow fibre filter modules, PES membrane, MiniKros®
Leverandør: Spectrum Laboratories
N02-P20U-10-N
SPECN02-P20U-10-NEA 23525 NOK
SPECN02-P20U-10-N
Hollow fibre filter modules, PES membrane, MiniKros®
Filter Systemer for tangentiell flow
Hollow fibre filter modules are particularly useful in tangential flow filtration because processes can easily be scaled-up from small volume applications to full-scale production.
  • Hollow fibre membranes are anisotropic in structure
  • The filter housings are engineered to handle higher pressures and autoclaving
  • Low protein binding increases the product yield of each processing run
  • TC filters have FLL/FLL connections and come in three effective lengths
Tangential flow filtration is used in both microfiltration and ultra-filtration applications. Ultra-More Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Inngangs-/utgangskobling forbindelser 1/2" Tri-clamp/3/4" Tri-clamp