ProConnex® Standard Flow Path for MidiKros or MidiKros TC

Leverandør: Spectrum Laboratories
STUBEGN15390S
SPECSTUBEGN15390SEA 68300 NOK
SPECSTUBEGN15390S
ProConnex® Standard Flow Path for MidiKros or MidiKros TC
Filter Systemer for tangentiell flow
ProConnex® standard flow path for midiKros or midiKros TC, 250 ml, 3/Pk.

System: KR2i; pressor sensors: luer; delivery condition: sterile; compatible with: MidiKros (D) or MidiKros TC (T).

HUV recirculation: 3,1 ml; HUV reservoir: 1 ml; HUV total tubeset: 34,7 ml.

Bestillingsinformasjon: Filter sold separately.
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR