ProConnex® Standard Flow Path for MiniKros Sampler
Leverandør: Spectrum Laboratories
STUBEGN15391S
SPECSTUBEGN15391SEA 44325 NOK
SPECSTUBEGN15391S
ProConnex® Standard Flow Path for MiniKros Sampler
Filter Systemer for tangentiell flow
ProConnex® standard flow path for MiniKros sampler, 1 L.
System: KR2i; pressor sensors: luer; delivery condition: sterile; compatible with: MiniKros sampler (S).
HUV recirculation: 19,9 ml; HUV reservoir: 19,3 ml; HUV total tubeset: 152,1 ml.
Bestill nå
SPESIFIKASJONER
Type Flatbottom