Cassette
Leverandør: Spectrum Laboratories
XP300MP1L
SPECXP300MP1LEA 0 NOK
SPECXP300MP1L
Cassette
Filter Kassetter for tangentiell flow
SIUS PD cassette with HyStream membrane type and molecular weight cut-off (MWCO) of 300kD.
  • medium channel
  • Single-use
  • Reusable