Cryopreservation media, HyClone™ HyCryo™
Leverandør: HyClone products (Cytiva)
SR30001.02
SR30001.02EA 2975 NOK
SR30001.02
Cryopreservation media, HyClone™ HyCryo™
Media, Cellelagring
Animal origin-free HyClone™ HyCryo cryopreservation media is designed for general use.
  • 0,2 μm sterile filtered
  • Serum free formulation
  • Increased recovery of viable cells versus standard cryopreservation formulations