Bovine Thrombin (from Plasma)

Leverandør: SIGMA ALDRICH MICROSCOPY

SRE0003-10KU
SRE0003-10KUEA 7375 NOK
SRE0003-10KU
Bovine Thrombin (from Plasma)
Enzymer

Serine protease that selectively cleaves Arg-Gly bonds in fibrinogen to form fibrin and fibrinopeptides A and B.


  • Activity is expressed in NIH units obtained by direct comparison to a NIH thrombin reference standard


The NIH assay procedure uses 0,2 ml of diluted plasma (1:1 with saline) as a substrate and 0,1 ml of thrombin sample (stabilised in a 1% buffered albumin solution) based on a modification of the method of Biggs. Only clotting times in the range of 15 to 25 seconds are used for determining thrombin concentrations.

Bestill nå

SPESIFIKASJONER

Enzymnavn Thrombin
Environmentally Preferable
Kilde Plasma
Arter Bovine
MDL No. MFCD00166053
CAS-nr 9002-04-4
Skjema Powder
Lagringsforhold −20 °C
Fraktforhold Wet ice
Enzymaktivitet 80 - 300 NIH units/mg protein
M-Clarity quality level MQ400

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR