Erythropoietin (EPO) ELISA Kit
Leverandør: STEMCELL Technologies
01630
STMC01630EA 0 NOK
STMC01630
Erythropoietin (EPO) ELISA Kit
Assay ELISAs
Immunoassay for detection and measurement of human erythropoietin.
The glycoprotein EPO is the main physiological regulator of red blood cell formation. The EPO ELISA kit is a rapid 3-step enzyme-linked immunosorbent assay (ELISA) designed for the quantitative measurement of natural and recombinant human EPO.
The assay uses two monoclonal antibodies raised against human urinary EPO. These antibodies bind two non-overlapping epitopes on the EPO polypeptide and show high-affinity binding to both natural and recombinant EPO. The range of detection for the kit is 1,6.
NB: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Mål 1630
Primary antibody reactivity Human
Vert