ELISA Diluent
Leverandør: STEMCELL Technologies
01916
STMC01916EA 0 NOK
STMC01916
ELISA Diluent
Assay ELISAs
Diluent for ELISA samples, standards, and detection antibodies.
ELISA diluent is intended for dilution of samples, standards, and detection antibodies in ELISA assays. By preventing the cross-linking of heterophilic antibodies, ELISA diluent minimises interference and false-positive signals in ELISA reactions. ELISA Diluent is required for use with the human IgE ELISA antibody pair kit when serum or plasma samples are used.
NB: Do not use ELISA diluent for dilution of streptavidin conjugates.
Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').