Human IgA ELISA Antibody Pair Kit
Leverandør: STEMCELL Technologies
01992
STMC01992EA 0 NOK
STMC01992
Human IgA ELISA Antibody Pair Kit
Assay ELISAs
For detection and measurement of human immunoglobulin A.
The Human Immunoglobulin A (IgA) ELISA antibody pair Kit is intended for those who want the flexibility of setting up their own ELISA assay. This kit includes capture and detection antibodies and an IgA standard. It is designed for the quantitative detection and measurement of total human IgA in biological fluids such as serum, plasma, and cell culture supernatants.
IgA is predominantly found on the surface of epithelial cells where it acts as a neutralising antibody against invading pathogens. IgA primarily exists as a dimer in secretions such as saliva, tears, colostrum, mucus, sweat, and gastric fluid, but also occurs as a monomer in serum, where it comprises 10.
NB: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Mål IgA
Primary antibody reactivity Human
Vert