Mouse IgM ELISA Antibody Pair Kit
Leverandør: STEMCELL Technologies
01999
STMC01999EA 0 NOK
STMC01999
Mouse IgM ELISA Antibody Pair Kit
Assay ELISAs
For detection and measurement of mouse immunoglobulin M.
The Mouse Immunoglobulin M (IgM) ELISA Antibody Pair Kit is intended for those who want the flexibility of setting up their own ELISA assay. This kit includes capture and detection antibodies and an IgM standard. It is designed for the quantitative detection and measurement of mouse IgM in biological fluids such as serum, plasma, and cell culture supernatants.
IgM is predominantly found in lymph fluid and in blood where it acts as a neutralizing agent in the early stages of infection. Secreted IgM exists in a pentameric form whereas monomeric IgM is found on the surface of B cells. IgM constitutes approximately 10% of all serum immunoglobulins. The assay is based on the sandwich ELISA method, in which samples are added to ELISA plates coated with capture antibodies specific for the immunoglobulin. The captured immunoglobulin is detected by addition of a biotinylated detection antibody, followed by streptavidin-alkaline phosphatase (SA-ALP), which binds the biotinylated antibody.
Addition of the chromogenic enzyme substrate p-nitrophenyl phosphate (pNPP) results in a coloured product with an intensity directly proportional to the concentration of immunoglobulin in the sample. The concentration of the immunoglobulin is determined by comparison to a serial dilution of the immunoglobulin standard analyzed in parallel.
Note: This kit includes sufficient reagents for 6×96-well ELISA plates. ELISA plates (Catalog #STMC038019) and pNPP Substrate (Catalog #STMC01917) are required for use with the Mouse IgM ELISA Antibody Pair Kit and are available for purchase separately.
NB: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').