Human IL-2 ELISA Kits
Leverandør: STEMCELL Technologies
02006 02007
STMC02006EA 0 NOK
STMC02006 STMC02007
Human IL-2 ELISA Kits
Assay ELISAs
For detection and measurement of human interleukin 2.
The Human Interleukin 2 (IL-2) ELISA Kit is designed for the quantitative detection and measurement of human IL-2 in biological fluids such as serum, plasma, and cell culture supernatants. IL-2 is primarily secreted by activated CD⁴⁺ and CD⁸⁺ T cells, but is also produced by B cells and dendritic cells in smaller amounts.
IL-2 is a major regulator of T cell development, contributing to T cell growth and expansion, as well as the development of regulatory T cells. The assay is based on the sandwich ELISA method, in which samples are added to ELISA strip plates pre-coated with capture antibodies specific for the cytokine.
The captured cytokine is detected by addition of a biotinylated detection antibody, followed by streptavidin-horseradish peroxidase, which binds the biotinylated antibody. Addition of the chromogenic enzyme substrate 3,3’,5,5’ tetramethylbenzidine (TMB) results in a coloured product with an intensity directly proportional to the concentration of cytokine in the sample.
The concentration of the cytokine is determined by comparison to a serial dilution of the cytokine standard analysed in parallel.
NB: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').