CD34+ Progenitor Reprogramming Kit, STEMCELL Technologies
Leverandør: STEMCELL Technologies
05925
STMC05925EA 0 NOK
STMC05925
CD34+ Progenitor Reprogramming Kit, STEMCELL Technologies
Media, Cellekultur
Integrated workflow containing tools to isolate and expand CD34+ progenitor cells from peripheral blood and their subsequent reprogramming to iPS cells.
The CD34+ progenitor reprogramming kit contains an integrated set of tools and reagents to enrich, expand and reprogram CD34+ progenitor cells from human peripheral blood samples.
NB: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').