Soybean Trypsin Inhibitor, ACF
Leverandør: STEMCELL Technologies
07458 07457
STMC07458EA 0 NOK
STMC07458 STMC07457
Soybean Trypsin Inhibitor, ACF
Cellekultursupplementer og tilsetninger
For inhibition of trypsin activity.
  • Keep cultures free of animal-derived materials with a soybean-sourced trypsin inhibitor
  • Effectively inhibits trypsin, as well as human and bovine thrombin, plasmin, and leukocytic proteases
Inactivate trypsin with Soybean Trypsin Inhibitor, Animal Component-Free (ACF). This trypsin inhibitor consists of a single polypeptide chain with two disulfide bridges. It is able to inhibit chymotrypsin to a lesser extent, but is ineffective against esterolytic, proteolytic, or elastolytic activities of porcine elastase (Vered et al.). Soybean Trypsin inhibitor, ACF has also been shown to inhibit human and bovine thrombin, plasmin, and leukocytic proteases.
NB: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').