OptiPrep™ Density Medium
Leverandør: STEMCELL Technologies
07820
STMC07820EA 0 NOK
STMC07820
OptiPrep™ Density Medium
Media, Celleseparasjon
Density gradient medium for the isolation of viruses, organelles, macromolecules, or cells.
  • Use to form continuous or discontinuous gradients
  • Customise separation characteristics for isolation of viruses, organelles, macromolecules, or cells
  • Non-ionic, non-toxic to cells, isoosmotic, and metabolically inert
Simplify the isolation and purification of macromolecules, viruses, a wide range of cell types, and organelles such as nuclei, mitochondria, endosomes, or exosomes by using OptiPrep™ medium. This flexible, versatile, and gentle density gradientMore Product Information