Spleen Dissociation Medium
Leverandør: STEMCELL Technologies
07915
STMC07915EA 0 NOK
STMC07915
Spleen Dissociation Medium
Cellekultursupplementer og tilsetninger
Medium for dissociation of mouse spleen.
  • Maximise recovery and viability of dendritic cells from mouse spleen
  • Optimised for use with selected EasySep™ cell separation kits
Maximise the recovery of dendritic cells from mouse spleens with Spleen dissociation medium, when used in combination with EasySep™ cell separation technology. This enzymatic dissociation medium contains collagenase, DNase, and fetal bovine serum (FBS), and has been optimised to maximise the viability of isolated spleen dendritic cells. Each 4 ml tube is sufficient for processing up to two mouse spleens.
NB: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').