STEMdiff™ Astrocyte Differentiation Kit
Leverandør: STEMCELL Technologies
100-0013
STMC100-0013EA 0 NOK
STMC100-0013
STEMdiff™ Astrocyte Differentiation Kit
Media, Cellekultur
Differentiation kit for the generation of astrocyte precursors from human ES and iPS cell-derived neural progenitor cells.
STEMdiff™ astrocyte differentiation kit is used to rapidly and efficiently generate astrocytic precursors from neural progenitor cells (NPCs) derived from human pluripotent stem cells (hPSCs) using STEMdiff™ SMADi neural induction kit. These astrocytic precursors are then matured further into astrocytes using STEMdiff™ astrocyte maturation kit. Using this system, a highly pure population of astrocytes (an average of >70% S100B-positive and >60% GFAP-positive astrocytes; >15% doublecortin-positive neurons) can be generated from hPSCs in as few as 7 weeks and can be maintained long term in culture. Cells derived using these products are versatile tools for modeling human neurological development and disease, drug screening, toxicity testing, and cell therapy validation.
NB: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').