Anti-CD9 Mouse Monoclonal Antibody [clone: HI9a]
Leverandør: STEMCELL Technologies
100-0138
STMC100-0138EA 0 NOK
STMC100-0138
Anti-CD9 Mouse Monoclonal Antibody [clone: HI9a]
Antistoffer
Mouse monoclonal antibody against human, rhesus, cynomolgus CD9, unconjugated.
The HI9a antibody reacts with an extracellular epitope on human CD9, a 24 kDa single-chain type III transmembrane glycoprotein. CD9 is broadly expressed, including on the surface of monocytes, developing B cells, activated T cells, granulocytes, epithelial cells, endothelial cells, and platelets (mainly the α-granules) and is also a marker characteristic of extracellular vesicles. CD9 belongs to the tetraspanin family of proteins, which contain four transmembrane domains, two extracellular loops, and shortMore Product Information